siRNA Details
Gene Symbol: FLJ45513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242791.1 | NP_001229720.1 | 3 | 1429 | ||
GenBank mRNA | AK127421.1 | - | 3 | 1425 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ45513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242791.1 | NP_001229720.1 | 3 | 1429 | ||
GenBank mRNA | AK127421.1 | - | 3 | 1425 |