siRNA Details
Gene Symbol: LOC101241902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077236.1 | 1 | 889 | |||
GenBank mRNA | BC072451.1 | - | 1 | 889 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101241902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077236.1 | 1 | 889 | |||
GenBank mRNA | BC072451.1 | - | 1 | 889 |