siRNA Details
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136519.1 | 3 | 1872 | |||
GenBank mRNA | AK092681.1 | - | 1 | 1543 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136519.1 | 3 | 1872 | |||
GenBank mRNA | AK092681.1 | - | 1 | 1543 |