siRNA Details
Gene Symbol: KCNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095539.1 | - | 2 | 878 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC019877.1 | - | 2 | 2048 | Chr. 2 - 95301687 - 95303890 [+] on Build GRCh38 |
