siRNA Details
Gene Symbol: LOC284080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038230.1 | 3 | 297 | |||
GenBank mRNA | AK096761.1 | - | 3 | 297 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA869944.1 | - | 3 | 297 | Chr. 17 - 50052348 - 50055739 [-] on Build GRCh38 |
