siRNA Details
Gene Symbol: LOC731157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105024.1 | 1 | 194 | |||
GenBank mRNA | AF161364.1 | - | 1 | 195 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF150372.1 | - | 1 | 195 | Chr. 5 - 90410000 - 90410668 [+] on Build GRCh38 |
