siRNA Details
Gene Symbol: LINGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258282.1 | NP_001245211.1 | 7 | 2908 | ||
NM_152570.2 | NP_689783.1 | 7 | 2988 | |||
XM_011517719.1 | XP_011516021.1 | 6 | 3856 | |||
XM_011517724.2 | XP_011516026.1 | 5 | 6875 | |||
XM_011517728.2 | XP_011516030.1 | 2 | 6654 | |||
XM_017014303.1 | XP_016869792.1 | 11 | 6759 | |||
XM_017014304.1 | XP_016869793.1 | 4 | 9279 | |||
XM_017014305.1 | XP_016869794.1 | 5 | 8862 | |||
XM_017014306.1 | XP_016869795.1 | 3 | 6023 | |||
XM_017014307.1 | XP_016869796.1 | 2 | 3496 |
