siRNA Details
Gene Symbol: LOC102724699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125409.1 | 2, 3 | 495 | |||
GenBank mRNA | AK094454.1 | - | 2, 3 | 495 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA072422.1 | - | 2, 3 | 495 | Chr. 3 - 186823545 - 186825521 [-] on Build GRCh38 |
