siRNA Details
Gene Symbol: ODF3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104468.1 | - | 3 | 1210 | 40032428 | |
BC104469.1 | - | 4 | 1314 | 40032432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL833870.1 | - | 2 | 590 | Chr. 19 - 463359 - 464607 [-] on Build GRCh38 |
