siRNA Details
Gene Symbol: TMCO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537504.2 | XP_011535806.1 | 9 | 2616 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126395.1 | - | 1 | 1869 | Chr. 13 - 113527100 - 113529168 [+] on Build GRCh38 |
