siRNA Details
Gene Symbol: LINC01111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105006.1 | 2 | 177 | |||
GenBank mRNA | BC015449.1 | - | 2 | 177 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG171060.1 | - | 2 | 176 | Chr. 8 - 76406655 - 76523428 [+] on Build GRCh38 |
