siRNA Details
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518172.2 | XP_011516474.1 | 33 | 5496 | ||
XM_017014227.1 | XP_016869716.1 | 32 | 5514 | |||
XM_017014228.1 | XP_016869717.1 | 32 | 5370 |
