siRNA Details
Gene Symbol: LOC644727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095971.1 | - | 1 | 2025 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG150888.1 | - | 1 | 218 | Chr. 8 - 52715436 - 52715813 [+] on Build GRCh38 |
