siRNA Details
Gene Symbol: LOC101927058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110042.1 | 1 | 32 | |||
GenBank mRNA | BC042591.1 | - | 1 | 32 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG719776.1 | - | 1 | 37 | Chr. 12 - 42629655 - 42646498 [-] on Build GRCh38 |
