siRNA Details
Gene Symbol: BLACE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103545.1 | 4 | 433 | |||
GenBank mRNA | AY166699.1 | - | 4 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM478687.1 | - | 134 | Chr. - - [-] on Build |
