siRNA Details
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 2 | 802 | |||
NR_046252.1 | 2 | 792 | ||||
NR_046253.1 | 2 | 769 | ||||
NR_046254.1 | 2 | 765 | ||||
NR_046255.1 | 2 | 528 | ||||
GenBank mRNA | AK055924.1 | - | 2 | 770 | ||
AK096279.1 | - | 2 | 785 | |||
BC008609.1 | - | 2 | 471 | |||
BC039246.1 | - | 2 | 752 | |||
BC056913.1 | - | 2 | 528 | |||
BC092452.1 | - | 2 | 533 |
