siRNA Details
Gene Symbol: SPATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093385.1 | - | 6 | 1094 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR595989.1 | - | 1 | 1997 | Chr. 2 - 231012058 - 231014422 [+] on Build GRCh38 |
