siRNA Details
Gene Symbol: GTF3C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012204.3 | NP_036336.2 | 5 | 4715 | ||
NR_133925.1 | 6 | 4753 | ||||
XM_017015318.1 | XP_016870807.1 | 5 | 2498 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GTF3C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012204.3 | NP_036336.2 | 5 | 4715 | ||
NR_133925.1 | 6 | 4753 | ||||
XM_017015318.1 | XP_016870807.1 | 5 | 2498 |