siRNA Details
Gene Symbol: NKTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533747.2 | XP_011532049.1 | 8 | 695 | ||
XM_017006476.1 | XP_016861965.1 | 2 | 1889 | |||
GenBank mRNA | AB209888.1 | - | 6 | 2614 | ||
BX538094.1 | - | 2662 | ||||
BX641127.1 | - | 1 | 205 |
