siRNA Details
Gene Symbol: SERTAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273193.4 | XP_005273250.1 | 4 | 700 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK021425.1 | - | 1 | 1408 | Chr. 1 - 210244422 - 210246253 [+] on Build GRCh38 |
