siRNA Details
Gene Symbol: RBFADN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF070628.1 | - | 1 | 1258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM930419.1 | - | 1 | 113 | Chr. 18 - 80096763 - 80097092 [+] on Build GRCh38 |
