siRNA Details
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA864573.1 | - | 1 | 250 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124606.1 | - | 1 | 2586 | Chr. 4 - 1164578 - 1168174 [+] on Build GRCh38 |
