siRNA Details
Gene Symbol: C1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270975.4 | XP_005271032.1 | 1 | 864 | ||
XM_005270981.4 | XP_005271038.1 | 1 | 864 | |||
XM_011541646.2 | XP_011539948.1 | 1 | 864 | |||
XM_017001546.1 | XP_016857035.1 | 1 | 863 | |||
GenBank mRNA | AK056029.1 | - | 1 | 1755 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX644786.1 | - | 1 | 128 | Chr. 1 - 37689953 - 37690708 [-] on Build GRCh38 |
