siRNA Details
Gene Symbol: HSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286503.1 | NP_001273432.1 | 17 | 4072 | ||
NM_001286504.1 | NP_001273433.1 | 18 | 4065 | |||
NM_001286505.1 | NP_001273434.1 | 16 | 3831 | |||
NM_006644.3 | NP_006635.2 | 18 | 4204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR611122.1 | - | 1 | 347 | Chr. 13 - 31135049 - 31136476 [-] on Build GRCh38 |
