siRNA Details
Gene Symbol: LOC101928131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121677.1 | 4 | 661 | |||
GenBank mRNA | BC039474.1 | - | 4 | 661 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM450007.1 | - | 4 | 660 | Chr. 4 - 165731364 - 165743070 [-] on Build GRCh38 |
