siRNA Details
Gene Symbol: LOC101928748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110950.1 | 2, 3 | 482 | |||
GenBank mRNA | BC034316.1 | - | 2, 3 | 482 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721486.1 | - | 2, 3 | 484 | Chr. 9 - 115127348 - 115138400 [-] on Build GRCh38 |
