siRNA Details
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021894.1 | - | 1 | 1846 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EB385780.1 | - | 1 | 766 | Chr. 4 - 113378749 - 113379573 [+] on Build GRCh38 |
