siRNA Details
Gene Symbol: LINC00970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104091.1 | 5 | 394 | |||
GenBank mRNA | BC041008.1 | - | 5 | 399 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI544897.1 | - | 5 | 399 | Chr. 1 - 168904133 - 169087007 [-] on Build GRCh38 |
