siRNA Details
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 4602 | ||
NM_001281711.1 | NP_001268640.1 | 18 | 4563 | |||
NM_001281712.1 | NP_001268641.1 | 17 | 4382 | |||
NM_015341.4 | NP_056156.2 | 18 | 4635 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR608905.1 | - | 1 | 1008 | Chr. 2 - 96374664 - 96377067 [+] on Build GRCh38 |
