siRNA Details
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 5344 | ||
NM_001281711.1 | NP_001268640.1 | 18 | 5305 | |||
NM_001281712.1 | NP_001268641.1 | 17 | 5124 | |||
NM_015341.4 | NP_056156.2 | 18 | 5377 |
