siRNA Details
Gene Symbol: LOC101929621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134679.1 | 2 | 398 | |||
GenBank mRNA | BC038206.1 | - | 2 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG392299.1 | - | 2 | 398 | Chr. 4 - 108557233 - 108560396 [+] on Build GRCh38 |
