siRNA Details
Gene Symbol: LOC387810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104155.1 | 4 | 499 | |||
GenBank mRNA | BC070394.1 | - | 4 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU506733.1 | - | 4 | 501 | Chr. 11 - 112535559 - 112555802 [-] on Build GRCh38 |
