siRNA Details
Gene Symbol: ZNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720164.3 | XP_006720227.1 | 1 | 71 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK056849.1 | - | 2 | 1073 | Chr. 14 - 21101704 - 21104216 [-] on Build GRCh38 |
