siRNA Details
Gene Symbol: LOC100132174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104134.1 | 1 | 50 | |||
GenBank mRNA | AK127897.1 | - | 1 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA925334.1 | - | 1 | 50 | Chr. 17 - 77724558 - 77728559 [-] on Build GRCh38 |
