siRNA Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024563.1 | XP_016880052.1 | 1 | 1211 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF799301.1 | - | 1 | 64 | Chr. 17 - 48583419 - 48583736 [-] on Build GRCh38 |
