siRNA Details
Gene Symbol: LOC101928241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110693.1 | 3 | 153 | |||
GenBank mRNA | AK056049.1 | - | 3 | 153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA684621.1 | - | 3 | 153 | Chr. 1 - 96255525 - 96374125 [-] on Build GRCh38 |
