siRNA Details
Gene Symbol: LINC01355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110616.1 | 2 | 2945 | |||
GenBank mRNA | AL832122.1 | - | 1 | 4878 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF677896.1 | - | 1 | 176 | Chr. 1 - 23281320 - 23282051 [-] on Build GRCh38 |
