siRNA Details
Gene Symbol: PDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020786.2 | NP_065837.1 | 2 | 6615 | ||
XM_005256064.4 | XP_005256121.1 | 2 | 6662 | |||
XM_005256066.4 | XP_005256123.1 | 2 | 6704 | |||
XM_005256067.4 | XP_005256124.1 | 2 | 6729 | |||
XM_005256068.4 | XP_005256125.1 | 3 | 6676 | |||
XM_017023506.1 | XP_016878995.1 | 3 | 6810 |
