siRNA Details
Gene Symbol: GS1-204I12.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508908.1 | - | 2 | 440 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE906931.1 | - | 3 | 468 | Chr. 1 - 185651167 - 185656997 [+] on Build GRCh38 |
