siRNA Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722145.3 | XP_006722208.1 | 4 | 4178 | ||
XM_017025247.1 | XP_016880736.1 | 4 | 4178 | |||
XM_017025248.1 | XP_016880737.1 | 3 | 4080 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC047422.1 | - | 1 | 840 | Chr. 17 - 75796209 - 75799085 [+] on Build GRCh38 |
