siRNA Details
Gene Symbol: LOC101929745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134272.1 | 1 | 99 | |||
GenBank mRNA | BC028978.1 | - | 1 | 99 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE892334.1 | - | 1 | 78 | Chr. 5 - 38148501 - 38151918 [+] on Build GRCh38 |
