siRNA Details
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL049423.1 | - | 1 | 938 | ||
BC035518.1 | - | 1 | 1397 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL049423.1 | - | 1 | 938 | ||
BC035518.1 | - | 1 | 1397 |