siRNA Details
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127173.1 | - | 4 | 1232 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK123289.1 | - | 2 | 791 | Chr. 10 - 122985833 - 122988218 [+] on Build GRCh38 |
