siRNA Details
Gene Symbol: LOC101929586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120363.1 | 3 | 500 | |||
GenBank mRNA | BC041477.1 | - | 3 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI914475.1 | - | 3 | 502 | Chr. 15 - 80556344 - 80562945 [-] on Build GRCh38 |
