siRNA Details
Gene Symbol: LOC101929694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110017.1 | 1, 2 | 83 | |||
GenBank mRNA | BC033333.1 | - | 1, 2 | 85 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG718999.1 | - | 1, 2 | 90 | Chr. 3 - 112740579 - 112749319 [-] on Build GRCh38 |
