siRNA Details
Gene Symbol: HHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263178.4 | XP_005263235.1 | 14 | 5608 | ||
XM_006714288.3 | XP_006714351.1 | 14 | 5604 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124396.1 | - | 1 | 2101 | Chr. 4 - 144739637 - 144745270 [+] on Build GRCh38 |
