siRNA Details
Gene Symbol: LOC283887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135196.1 | 2 | 1200 | |||
NR_135197.1 | 2 | 1186 | ||||
GenBank mRNA | AK093382.1 | - | 2 | 1049 | ||
BC023651.2 | - | 2 | 1136 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135196.1 | 2 | 1200 | |||
NR_135197.1 | 2 | 1186 | ||||
GenBank mRNA | AK093382.1 | - | 2 | 1049 | ||
BC023651.2 | - | 2 | 1136 |