siRNA Details
Gene Symbol: TTLL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251728.2 | XP_005251785.1 | 8 | 7681 | ||
XM_005251729.4 | XP_005251786.1 | 9 | 8758 | |||
GenBank mRNA | AK123875.1 | - | 1 | 1691 | ||
BC040664.2 | - | 1 | 910 |
