siRNA Details
Gene Symbol: ZKSCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242894.1 | NP_001229823.1 | 7 | 4574 | ||
NM_001242895.1 | NP_001229824.1 | 5 | 3993 | |||
NM_024493.3 | NP_077819.2 | 6 | 4457 | |||
XM_005249423.3 | XP_005249480.1 | 5 | 4623 | |||
XM_006715215.2 | XP_006715278.1 | 6 | 4355 | |||
XM_006715216.2 | XP_006715279.1 | 6 | 4370 | |||
XM_006715218.3 | XP_006715281.1 | 5 | 3921 | |||
XM_017011320.1 | XP_016866809.1 | 7 | 4451 |
