siRNA Details
Gene Symbol: LINC01282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110385.1 | 4 | 580 | |||
GenBank mRNA | BC043536.1 | - | 4 | 580 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI828357.1 | - | 4 | 580 | Chr. X - 39367512 - 39391774 [-] on Build GRCh38 |
